Pith Ball Electroscopes | Avantor
Pith Ball Electroscopes
Pith ball electroscope
Catalog # 470336-558
Supplier:   American Scientific
Fournisseur #:  3134-01
Pith Ball Electroscopes
Catalog # 470336-558
Pith Ball Electroscopes
Pith Ball Electroscopes
Pith ball electroscope
Price
$10.05
Log in pour la disponibilité
Each
Product Details & Documents
Simple, timeless demonstration of electrostatics.
  • Comes with a teacher’s guide
  • Additional pith balls available in packs of 10
Use this item for demonstrating the attraction and repulsion of electric charges. It consists of an insulated brass pillar and hook on a base. It is useful for interesting and engaging demonstrations.
Delivery information: Supplied with two colored pith balls with strings attached.
Spécifications
  • Désignation:
    Pith ball electroscope