Pith Ball Electroscopes
Pith ball electroscope
Catalog # 470336-558
Supplier: American Scientific
Fournisseur #: 3134-01
Jump to:
Product Details & Documents
Simple, timeless demonstration of electrostatics.
- Comes with a teacher’s guide
- Additional pith balls available in packs of 10
Use this item for demonstrating the attraction and repulsion of electric charges. It consists of an insulated brass pillar and hook on a base. It is useful for interesting and engaging demonstrations.
Delivery information: Supplied with two colored pith balls with strings attached.
Spécifications
- Désignation:Pith ball electroscope