VWR® Cell Holders
Cell holder, 1 position, thermostattable, for 10 mm standard cells (external water bath required)
Catalog # 76520-722
Supplier: VWR International
Jump to:
Product Details & Documents
Specifications
- Désignation:Cell holder, 1 position, thermostattable, for 10 mm standard cells (external water bath required)