SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
Bond Elut cartridge, Cartouche, SPEC C18AR/MP3, 10 ml, 70 mg
  CAAGA5022570
Supplier #:  A5022570
SPEC Disk SPE, Agilent Technologies
  CAAGA5022570
SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
Bond Elut cartridge, Cartouche, SPEC C18AR/MP3, 10 ml, 70 mg
$901.00
Pack of 100
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
  • Type de colonne:
    Cartouche
  • Capacité:
    10 ml
  • Désignation:
    Bond Elut cartridge
  • Bed Weight:
    70 mg
  • Phase:
    SPEC C18AR/MP3