CrossLab Autosampler Sampling Capillaries, Agilent Technologies | Avantor
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Capillary assembly, PTFE, 0.022 in Int. Ø, 0.05" Ext. Ø
Catalog # CAAG9910115100
Fournisseur #:  9910115100
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Catalog # CAAG9910115100
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Capillary assembly, PTFE, 0.022 in Int. Ø, 0.05" Ext. Ø
Price
$483.00
Log in pour la disponibilité
Pack of 5
Product Details & Documents
Dispensing capillary assemblies forAgilent PSD 100/120 Programmable Sample Dispenser.
  • Works well with solvent and acid-resistant sampling components
  • PTFE capillary tips and capillaries reduce contamination
  • Easy replacement by the user
Agilent provides a range of supplies for the PSD 100/120 Programmable Sample Dspenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.​
Dispensing capillary assemblies are made from PTFE tubing, making them ideal for collecting and dispensing a variety of corrosive matrices and organic solutions.
Spécifications
  • Désignation:
    Capillary assembly, PTFE, 0.022 in Int. Ø, 0.05" Ext. Ø