CrossLab Dissolution Shafts, Agilent Technologies
CrossLab Dissolution Shafts, Agilent Technologies
Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, USP, Pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
  CAAG16-3631
Supplier #:  16-3631
CrossLab Dissolution Shafts, Agilent Technologies
  CAAG16-3631
CrossLab Dissolution Shafts, Agilent Technologies
CrossLab Dissolution Shafts, Agilent Technologies
Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, USP, Pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
$2,180.00
Each
Shafts of different materials and lengths, individually serialized to meet standard USP design.
  • Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
  • Individually serialized
  • Three-clip shafts to meet standard USP design
  • O-ring shafts for automated systems
  • Conical shafts to support air bubble dispersion
  • A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
  • Désignation:
    Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, USP, Pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
  • Certification:
    Yes