CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Screw, stainless steel, bead adjuster, locking, M8, for Mark 7 spray chamber
Catalog # CAAG1510226000
Supplier #:  1510226000
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Catalog # CAAG1510226000
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
CrossLab Spray Chamber Components for Atomic Absorption Instruments, Agilent Technologies
Screw, stainless steel, bead adjuster, locking, M8, for Mark 7 spray chamber
Price
$37.10
Log in pour la disponibilité
Each
Product Details & Documents
Mark 7 atomization system for current model Agilent AA instruments.
  • Simple-to-use twist and lock assembly makes for easy cleaning and routine maintenance
  • Flexible operation with the capability to tune performance using the externally adjustable impact bead, fit mixing paddles for better precision, or both
  • Inert construction makes spray chamber suitable for use with aqueous and organic solutions
  • Fit organic resistant O-rings to ensure organic compatibility
The nebulizer, spray chamber and burner, plays an important part in controlling the analytical performance of a flame AA spectrometer. Proper spray chamber design ensures the minimization of carbon build-up, burner blockage and volatilization interferences. The spray chamber must also provide excellent drainage.
The Mark 7 spray chamber features an adjustable impact bead that breaks up aerosol droplets and provides the operator with control over the aerosol concentration. A set of removable twin-head mixing paddles promote mixing and large droplet removal. These features provide a smaller drop size aerosol, reducing burner blockage, signal noise, and improving long term signal stability.
Spécifications
  • Désignation:
    Screw, stainless steel, bead adjuster, locking, M8, for Mark 7 spray chamber