CrossLab Dissolution Shafts, Agilent Technologies | Avantor
CrossLab Dissolution Shafts, Agilent Technologies
Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 14.75", for 705-DS; 708-DS; 709-DS; VK7000
Catalog # CAAG14-3613
Fournisseur #:  14-3613
CrossLab Dissolution Shafts, Agilent Technologies
Catalog # CAAG14-3613
CrossLab Dissolution Shafts, Agilent Technologies
CrossLab Dissolution Shafts, Agilent Technologies
Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 14.75", for 705-DS; 708-DS; 709-DS; VK7000
Price
379,00 $
Log in pour la disponibilité
Each
Product Details & Documents
Shafts of different materials and lengths, individually serialized to meet standard USP design.
  • Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
  • Individually serialized
  • Three-clip shafts to meet standard USP design
  • O-ring shafts for automated systems
  • Conical shafts to support air bubble dispersion
  • A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
Spécifications
  • Désignation:
    Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 14.75", for 705-DS; 708-DS; 709-DS; VK7000
  • Certification:
    Yes