CrossLab Dissolution Paddles, Agilent Technologies | Avantor
CrossLab Dissolution Paddles, Agilent Technologies
Paddle, electropolished stainless steel, 21", for 705-DS; 708-DS; 709-DS; VK7000
Catalog # CAAG14-3595
Fournisseur #:  14-3595
CrossLab Dissolution Paddles, Agilent Technologies
Catalog # CAAG14-3595
CrossLab Dissolution Paddles, Agilent Technologies
CrossLab Dissolution Paddles, Agilent Technologies
Paddle, electropolished stainless steel, 21", for 705-DS; 708-DS; 709-DS; VK7000
Price
$533.00
Log in pour la disponibilité
Each
Product Details & Documents
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
  • All paddles are serialized and designed to meet USP specifications for Apparatus 2
  • Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
  • Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
Certifications: Each paddle includes an individual Certificate of Conformance as standard.
Spécifications
  • Désignation:
    Paddle, electropolished stainless steel, 21", for 705-DS; 708-DS; 709-DS; VK7000
  • Certification:
    Yes
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.