CrossLab Dissolution Flow Cells, Agilent Technologies
Flow Cell, 1 mm pathlength, for Cary 50/60
  CAAG11-1315
CrossLab Dissolution Flow Cells, Agilent Technologies
  CAAG11-1315
CrossLab Dissolution Flow Cells, Agilent Technologies
CrossLab Dissolution Flow Cells, Agilent Technologies
Flow Cell, 1 mm pathlength, for Cary 50/60
$1,860.00
Each
Flow cells for the Cary UV-Vis spectrophotometers integrate dissolution testing with online UV.
  • The Cary 60 makes it easy to interconnect two different cells within the multicell transport
  • Switch between two sets of cells, typically with different path lengths, with no disassembling or reassembling of the tubing lines
Flow cells for the Agilent Cary UV-visible spectrophotometer help integrate dissolution testing with online UV analysis to provide a single-source, automated, performance-testing solution.
  • Désignation:
    Flow Cell, 1 mm pathlength, for Cary 50/60