CrossLab Dissolution Flow Cells, Agilent Technologies
Flow Cell, 2 mm pathlength, for Cary 50/60
CAAG11-1310
: 11-1310
CrossLab Dissolution Flow Cells, Agilent Technologies
CAAG11-1310
Flow cells for the Cary UV-Vis spectrophotometers integrate dissolution testing with online UV.
- The Cary 60 makes it easy to interconnect two different cells within the multicell transport
- Switch between two sets of cells, typically with different path lengths, with no disassembling or reassembling of the tubing lines
Flow cells for the Agilent Cary UV-visible spectrophotometer help integrate dissolution testing with online UV analysis to provide a single-source, automated, performance-testing solution.
- Désignation:Flow Cell, 2 mm pathlength, for Cary 50/60