PIPETMAX® Software, Gilson | Avantor
PIPETMAX® Software, Gilson
Licence, PROTOCOL BUILDER V1, lifetime
Catalog # 76518-598
Supplier:   GILSON, INC.
Fournisseur #:  32000332
PIPETMAX® Software, Gilson
Catalog # 76518-598
Supplier:  GILSON, INC.
PIPETMAX® Software, Gilson
PIPETMAX® Software, Gilson
Licence, PROTOCOL BUILDER V1, lifetime
Price
$4,036.58
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
TRILUTION® micro Software manages your automated PIPETMAX® workflows, allowing you to focus on more important tasks.
  • Easy to learn
  • Improved reproducibility and traceability for PCR and qPCR experiments
  • No more tedious normailization calculations
TRILUTION micro Software, the runtime control software for PIPETMAX, takes only minutes to learn, unlike other automated sample preparation solutions that require extensive training, or even dedicated technicians. From a single screen, TRILUTION micro lets you:
• Manage protocol managements, including importing, exporting, and deleting protocols
• Simulate and run protocols
• Manually control processes
With the latest version 3.0, TRILUTION micro Software streamlines steps to rack editing and running protocols. TRILUTION micro Software v3.0 is available for Windows® 10 operating systems.
The qPCR Assistant add-on allows you to create PCR/qPCR methods that support sample dilution, master mix preparation and plate preparation. By automating these steps using PIPETMAX, you increase accuracy and reliability by minimizing variability and contamination sources. With the qPCR Assistant, it's easy to import sample information and export templates to different thermocyclers, adding a new level traceability of your experiments.
With the Normalization Assistant add-on, anyone in the lab can create fully automated DNA or RNA normalization protocols on PIPETMAX. Normalization Assistant automates the tedious calculations and sample tracking tasks required for normalization runs, improving traceability and reliability to your sample preparations.
Spécifications
  • Désignation:
    Licence, PROTOCOL BUILDER V1, lifetime