CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Lower cap, wider diameter for improved mixing, VanKel Type, for 300 ml tubes, for BIO-DIS
Catalog # AG27-1050
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
: 27-1050
CrossLab Reciprocating Dissolution Tubes, Agilent Technologies
Catalog # AG27-1050
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Several tube vessels are available for Apparatus 3 and 7.
- Reciprocating Cylinders for BIO-DIS and 400-DS
Ideal for automatic dissolution testing.
Spécifications
- Désignation:Lower cap, wider diameter for improved mixing, VanKel Type, for 300 ml tubes, for BIO-DIS