SPEC Disk SPE, Agilent Technologies
SPEC disks, C18AR, 47 mm, 47 mm
  CAA74819
SPEC Disk SPE, Agilent Technologies
  CAA74819
SPEC Disk SPE, Agilent Technologies
SPEC Disk SPE, Agilent Technologies
SPEC disks, C18AR, 47 mm, 47 mm
$305.00
Pack of 20
SPEC disk SPE delivers superior flow characteristics and trouble-free automation.
  • No loose sorbent means no channeling and better reproducibility
  • Uniform flow and extraction properties offer robust performance
  • Low elution volume affords excellent concentration of the analyte, improving sensitivity
SPEC disk SPE is a product providing faster sample processing, lower costs, and high-quality results. SPEC is manufactured with ultraclean polypropylene plastic and contains a monolithic sorbent bed with bonded silica infused on a glass fiber disk. For some applications, the elution volume is very small, eliminating the dry down and reconstitution steps.
Integrated filtration protects against clogging. SPEC is available in cartridge, 96-well plate, and disk formats in a wide range of chemistries for many applications.
  • Ø:
    47 mm
  • Désignation:
    SPEC disks, C18AR, 47 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.