CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25
Catalog # CA27-2005
Supplier #:  27-2005
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Catalog # CA27-2005
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25
Price
$392.00
Log in pour la disponibilité
Pack of 25
Product Details & Documents
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
  • Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Spécifications
  • Désignation:
    Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25