CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25
Catalog # CA27-2005
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Supplier #: 27-2005
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Catalog # CA27-2005
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Several reciprocating vessel screens are available for the BIO-DIS reciprocating cylinder apparatus.
- Reciprocating cylinder screens for BIO-DIS in stainless steel and polypropylene are available in different mesh sizes
Several reciprocating vessel screens are available for the Agilent BIO-DIS reciprocating cylinder apparatus, which is ideal for automatic dissolution testing of dosage forms that require different types of media.
Spécifications
- Désignation:Polypropylene screens, 1.25 inch Ext. Ø for 300 ml tubes, 40-mesh, 405 µm, for BIO-DIS, Pack of 25