Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
epT.I.P.S.® Sets, Pipette Tips | Avantor

epT.I.P.S.® Sets, Pipette Tips

0.5 - 20 µl, 46 mm, Light grey, 480 Tips (5 trays × 96 Tips), 1 reusable box
Catalog # 76516-706
   Low Manufacturing Impact
Fournisseur #:  0030076311
epT.I.P.S.® Sets, Pipette Tips
Catalog # 76516-706
Supplier:  
   Low Manufacturing Impact
epT.I.P.S.® Sets, Pipette Tips
epT.I.P.S.® Sets, Pipette Tips

0.5 - 20 µl, 46 mm, Light grey, 480 Tips (5 trays × 96 Tips), 1 reusable box

Discontinued
Product Details & Documents

Eppendorf epT.I.P.S.® Sets, in 'Eppendorf Quality' grade, combine the convenience of a reusable Box 2.0 with high-quality reloadable pipette tips, offering a sustainable, efficient solution for precise and organized pipetting in laboratories.

  • Contamination-free transfer of trays to stable working box
  • System optimized for use with multichannel pipettes
  • Color-coded trays for simple identification of tips and pipette
  • Tips can be attached to pipette from refill trays
  • Reloads and Boxes can be autoclaved at 121 °C for subsequent use
  • epT.I.P.S.® Box 2.0 and epT.I.P.S.® Set available in Eppendorf Quality
  • ACT® labeled by non-profit My Green Lab® to help make an informed choice regarding sustainability

The perfect fit for Eppendorf pipettes: epT.I.P.S.® ensure a reliable pipetting system:
Make everyday work easier and more enjoyable with the epT.I.P.S.® (Eppendorf 'Totally Integrated Pipetting System') pipette tips. These pipette tips are manufactured to extremely high quality and purity standards and are designed to cover all types of specific sampling needs. Offering reduced-force tip attachment and ejection with complete sealing, epT.I.P.S.® work in perfect combination with Eppendorf pipettes and form a fully integrated pipetting system that delivers maximum precision, accuracy, and reproducibility. Eppendorf pipette tips with ideal wetting properties and optimum transparency are manufactured exclusively from high-purity plastic materials. The universal nose cone design allows Eppendorf tips to also be used with pipettes from other manufacturers.

How to recognize an original Eppendorf pipette tip?
All Eppendorf tips feature a 'ring' of drop-shaped relief elements on the edge of the tip crown.

Extended length for reliable sampling: epT.I.P.S.® L pipette tips:
The elongated pipette tips (epT.I.P.S.® L) enable reliable sample reach while pipetting from and into conical tubes, high reagent bottles, narrow deep vessels, cell culture flasks, or deepwell plates. The long and slim design of these elongated pipette tips allows free access to the sample with reduced risk of touching the sides of tubes or wells. Cross-contamination during pipetting can be reduced to a minimum. epT.I.P.S.® L are available in volumes of: 0.5 to 20 µl, 50 to 1250 µl, 0.2 to 5 ml, and 0.5 to 10 ml.

 : General quality and ISO certificates available.

Spécifications
  • Volume:
    0.5 - 20 µl
  • Longueur:
    46 mm
  • Packed:
    480 Tips (5 trays × 96 Tips), 1 reusable box
  • Color Code:
    Light grey
  • Cat. No.:
    76516-706
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Low Manufacturing Impact