CrossLab Dissolution Flow Cells, Agilent Technologies
Flow Cell, 10 mm pathlength, for Cary 50/60
Catalog # CA11-1300
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Fournisseur #: 11-1300
CrossLab Dissolution Flow Cells, Agilent Technologies
Catalog # CA11-1300
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Jump to:
Product Details & Documents
Flow cells for the Cary UV-Vis spectrophotometers integrate dissolution testing with online UV.
- The Cary 60 makes it easy to interconnect two different cells within the multicell transport
- Switch between two sets of cells, typically with different path lengths, with no disassembling or reassembling of the tubing lines
Flow cells for the Agilent Cary UV-visible spectrophotometer help integrate dissolution testing with online UV analysis to provide a single-source, automated, performance-testing solution.
Spécifications
- Désignation:Flow Cell, 10 mm pathlength, for Cary 50/60