Slide-Sette Savers, Caplugs
High-impact polystyrene base, polypropylene saver, 20 Slides, White
Catalog # 76502-838
Supplier: Caplugs
Supplier #: 240-3828-W80
Jump to:
Product Details & Documents
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
- Each Slide-Sette Saver can be used to store specimens on a long-term basis
- Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
- Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
Specifications
- Capacité:20 Slides
- Color:White
- Matière:High-impact polystyrene base, polypropylene saver