Slide-Sette Savers, Caplugs
Slide-Sette Savers, Caplugs
High-impact polystyrene base, polypropylene saver, 20 Slides, White
Catalog # 76502-838
Supplier:   Caplugs
Supplier #:  240-3828-W80
Slide-Sette Savers, Caplugs
Catalog # 76502-838
Supplier:  Caplugs
Slide-Sette Savers, Caplugs
Slide-Sette Savers, Caplugs
High-impact polystyrene base, polypropylene saver, 20 Slides, White
Price
$76.66
Log in for availability
Pack of 3
Product Details & Documents
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
  • Each Slide-Sette Saver can be used to store specimens on a long-term basis
  • Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
  • Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
Specifications
  • Capacité:
    20 Slides
  • Color:
    White
  • Matière:
    High-impact polystyrene base, polypropylene saver