Slide-Sette Savers, Caplugs | Avantor
Slide-Sette Savers, Caplugs
PP, 1 slide, Yellow
Catalog # 76502-834
Supplier:   Caplugs
Fournisseur #:  240-3800-Y30
Slide-Sette Savers, Caplugs
Catalog # 76502-834
Supplier:  Caplugs
Slide-Sette Savers, Caplugs
Slide-Sette Savers, Caplugs
PP, 1 slide, Yellow
Price
$66.18
Log in pour la disponibilité
Bag of 100
Product Details & Documents
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
  • Each Slide-Sette Saver can be used to store specimens on a long-term basis
  • Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
  • Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
Spécifications
  • Capacité:
    1 slide
  • Color:
    Yellow
  • Matière:
    PP