Slide-Sette Savers, Caplugs
Slide-Sette Savers, Caplugs
PP, 1 slide, Natural
Catalog # 76502-822
Supplier:   Caplugs
Supplier #:  240-3800-030
Slide-Sette Savers, Caplugs
Catalog # 76502-822
Supplier:  Caplugs
Slide-Sette Savers, Caplugs
Slide-Sette Savers, Caplugs
PP, 1 slide, Natural
Price
$69.32
Log in for availability
Bag of 100
Product Details & Documents
The Slide-Sette Saver is designed for shipping same patient pathology microscope slides and tissue/biopsy paraffin blocks in a single container.
  • Each Slide-Sette Saver can be used to store specimens on a long-term basis
  • Eliminate time spent looking for misplaced slides and/or paraffin blocks when the need for retrieval arises
  • Available in multiple colors for various types of malignant tissue
The Slide-Sette Saver Storage Box is designed to hold multiple Slide-Sette Savers. It features a transparent polypropylene lid with a hinged bottom made of high-impact polystyrene.
Specifications
  • Capacité:
    1 slide
  • Color:
    Natural
  • Matière:
    PP