0,00 $
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Canada
ContactUsLinkComponent
FILTER MAT F/PRESTO A80 TEMP CNTRL SYS
CA89422-578
:
JML SCIENCES INC CA
FILTER MAT F/PRESTO A80 TEMP CNTRL SYS
CA89422-578
:
JML SCIENCES INC CA
:
8970921
Frequently Bought Together
Falcon® Centrifuge Tubes, Conical Bottom, Corning
: Corning
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
Acétonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH