Sticky Swabs, Electron Microscopy Sciences | Avantor
Sticky Swabs, Electron Microscopy Sciences
Sticky swabs, 3 mm
Catalog # 76499-604
Fournisseur #:  78324-65
Sticky Swabs, Electron Microscopy Sciences
Catalog # 76499-604
Sticky Swabs, Electron Microscopy Sciences
Sticky Swabs, Electron Microscopy Sciences
Sticky swabs, 3 mm
Price
$54.06
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Dust-free sticky swabs are specially used and designed for a high degree dust-free environment.
  • ESD safe
  • Made of PVC
  • Available in three sizes: 1, 2, and 3 mm
Sticky swabs have an anti-static treatment that can safely dissipate electrostatic charges. They are applicable for manufacturing and assembling static-sensitive products.
Packaging: Kit contains 25 of each size.
Spécifications
  • Désignation:
    Sticky swabs, 3 mm