Simulaids® Alex Patient Simulators | Avantor
Simulaids® Alex Patient Simulators
Alex, medium skin
Catalog # 470335-224
Supplier:   Nasco
Fournisseur #:  101-7130M
Simulaids® Alex Patient Simulators
Catalog # 470335-224
Supplier:  Nasco
Simulaids® Alex Patient Simulators
Simulaids® Alex Patient Simulators
Alex, medium skin
Price
$46,087.49
Log in pour la disponibilité
Each
Product Details & Documents
Sees, listens, and responds.
  • Three levels of options
  • Iris cam for patient perspective
  • One year warranty
The first patient communication simulator that sees, listens, and responds using artificial intelligence (AI). The Alex is an untethered, on-the-go patient simulator. Simply unpack, and you’re ready to train anytime and anywhere (Wi-Fi required). Speaks and understands English, French, and Spanish. Alex learns new languages all the time so it can help train more learners all around the globe.
Ordering information: These items have a limited return window. Please contact a sales rep if you want a preview or demonstration.
Spécifications
  • Désignation:
    Alex, medium skin