Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Catalog # 470335-412
Supplier:   LaMotte
 :  3354-CN-01
Nitrate-Nitrogen
Catalog # 470335-412
Supplier:  LaMotte
Nitrate-Nitrogen
Nitrate-Nitrogen
LaMotte® Water Test Kit, Nitrate-Nitrogen
Price
$134.45
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
  • Range: 0 to 15 ppm
  • Material for 50 tests
  • Compact, waterproof carrying case contains kit
  • Instructions included
  • Pre-calibrated test tubes
  • Aligned with GLOBE protocols
Spécifications
  • Désignation:
    LaMotte® Water Test Kit, Nitrate-Nitrogen