Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dilution Vials, Caplugs

Dilution Vials, Caplugs

25.00 ml, PS, Natural, Oui, LDPE, Natural, Bulk pack
Catalog # 76498-840
Supplier:   Caplugs
Supplier #:  201-5266-05K
Dilution Vials, Caplugs
Catalog # 76498-840
Supplier:  Caplugs
Dilution Vials, Caplugs
Dilution Vials, Caplugs

25.00 ml, PS, Natural, Oui, LDPE, Natural, Bulk pack

Price
$79.67
Log in for availability
Carton of 144
Product Details & Documents

Dilution vials provide accurate hemolysis and distortion-free cell and particle counts.

  • Provided in natural color
  • One-piece polystyrene design includes rounded seams, a frosted writing area and optically clear parallel sides for optimum light transmission
  • Leakproof caps have convenient tabs for easy removal

Vials are available in cases of 5×50 tray packs or in bulk pack of 144.

Specifications
  • Avec bouchon:
    Oui
  • Cap Material:
    LDPE
  • Capacité:
    25.00 ml
  • Color:
    Natural
  • Matière:
    PS
  • Packaging:
    Bulk pack
  • Cap Color:
    Natural
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.