Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Dilution Vials, Caplugs | Avantor

Dilution Vials, Caplugs

25.00 ml, PS, Natural, Oui, LDPE, Natural, 5×50 tray packs
Catalog # 76498-838
Supplier:   Caplugs
Fournisseur #:  201-5266-050
Dilution Vials, Caplugs
Catalog # 76498-838
Supplier:  Caplugs
Dilution Vials, Caplugs
Dilution Vials, Caplugs

25.00 ml, PS, Natural, Oui, LDPE, Natural, 5×50 tray packs

Price
$152.48
Log in pour la disponibilité
Carton of 250
Product Details & Documents

Dilution vials provide accurate hemolysis and distortion-free cell and particle counts.

  • Provided in natural color
  • One-piece polystyrene design includes rounded seams, a frosted writing area and optically clear parallel sides for optimum light transmission
  • Leakproof caps have convenient tabs for easy removal

Vials are available in cases of 5×50 tray packs or in bulk pack of 144.

Spécifications
  • Avec bouchon:
    Oui
  • Cap Material:
    LDPE
  • Capacité:
    25.00 ml
  • Color:
    Natural
  • Matière:
    PS
  • Packaging:
    5×50 tray packs
  • Cap Color:
    Natural