Paraffin Tapper | Avantor
Paraffin Tapper
Stainless steel paraffin tapper, 68×46×1.5 mm
Catalog # 76498-588
Supplier:   Mopec
Fournisseur #:  BG034
Paraffin Tapper
Catalog # 76498-588
Supplier:  Mopec
Paraffin Tapper
Paraffin Tapper
Stainless steel paraffin tapper, 68×46×1.5 mm
Price
$148.80
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to flatten tissue during the histology embedding process.
  • Stainless steel construction for durability and corrosion resistance
  • Ergonomic handle for comfortable, precise operation
  • Easy to clean and maintain for long-term use
  • Essential tool for pathology and histology workflows
Constructed from durable, laboratory-grade stainless steel, the Mopec Paraffin Tapper offers long-lasting performance and resistance to corrosion. Its ergonomic handle ensures a comfortable, controlled grip, reducing hand fatigue during repetitive use. The balanced design provides excellent control, making it ideal for use in histology, pathology, and research laboratories.
Spécifications
  • Désignation:
    Stainless steel paraffin tapper, 68×46×1.5 mm