Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Storage Containers with Optional Lids, Mortech® | Avantor

Storage Containers with Optional Lids, Mortech®

—, Storage container lid, 4.25 qt. container
Catalog # 76498-188
Supplier:   Mopec
Fournisseur #:  BA023
Storage Containers with Optional Lids, Mortech®
Catalog # 76498-188
Supplier:  Mopec
Storage Containers with Optional Lids, Mortech®
Storage Containers with Optional Lids, Mortech®

—, Storage container lid, 4.25 qt. container

Price
$157.32
Log in pour la disponibilité
Each
Product Details & Documents

Smooth stainless steel finish for easy cleaning.

  • All stainless steel, seamless construction
  • Smooth finish for easy cleaning
  • No pour spout
Spécifications
  • Capacité:
  • Désignation:
    Storage container lid
  • For Use With:
    4.25 qt. container