Storage Containers with Optional Lids, Mortech®
—, Storage container lid, 1.25 qt. container
Catalog # 76498-184
Supplier: Mopec
Supplier #: BA021
Jump to:
Product Details & Documents
Smooth stainless steel finish for easy cleaning.
- All stainless steel, seamless construction
- Smooth finish for easy cleaning
- No pour spout
Specifications
- Capacité:—
- Désignation:Storage container lid
- For Use With:1.25 qt. container



