Coplin Staining Jar, Mortech®
Coplin Staining Jar, Mortech®
Staining jar with plastic lid, 110H×16Ø mm
Catalog # 76498-154
Supplier:   Mopec
Supplier #:  BA003
Coplin Staining Jar, Mortech®
Catalog # 76498-154
Supplier:  Mopec
Coplin Staining Jar, Mortech®
Coplin Staining Jar, Mortech®
Staining jar with plastic lid, 110H×16Ø mm
Price
$71.29
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Heavy, clear glass jar with four molded internal grooves to hold up to ten slides.
  • Glass base construction
  • Four molded grooves in glass hold up to ten 3×1" slides
Jar features a broad base to prevent spillage. Both glass and plastic lid options protect against contaminants. The plastic option is a tight-fitting screw lid to prevent fluid evaporation. The glass cover option simply and easily sets upon the jar.
Specifications
  • Désignation:
    Staining jar with plastic lid
  • Dimensions:
    110H×16Ø mm