MINI CAPSULE HALF SIZE 1.2UM PP CS5
Catalogue # 76497-450
Fournisseur: PARKER HANNIFIN TE
MINI CAPSULE HALF SIZE 1.2UM PP CS5
Catalogue # 76497-450
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 22-CHBJJ-012-2
Frequently Bought Together
VWR® Traceable® Conductivity Calibration Standards (CRM)
Fournisseur: VWR International
Orion™ pH Buffer Individual Use Pouches, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
LOCTITE® 272 Thread lockers, Henkel Corporation
Fournisseur: Loctite
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Fournisseur: VWR International
MBP DNA AWAY™
Fournisseur: Thermo Scientific
TISSUE MARKING DYE - RD 2 OZ
Fournisseur: Electron Microscopy Sciences
BulletKit® SmGM®- 2 Smooth Muscle Cell Growth Medium -2
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Lactobacilli MRS Agar, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Fournisseur: Sekisui Diagnostics
AcroPrep™ Filter Plates for Aqueous Filtration, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
Tube Rocker, Electron Microscopy Sciences
Fournisseur: ELECTRON MICROSCOPY SCIENCE
Multi-Well Plate Vacuum Manifold, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
High Sensitivity DNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Puromycin dihydrochloride ≥98% (by HPLC)
Fournisseur: Enzo Life Sciences
Gastight® 1000 Series SampleLock™ Purge and Trap Syringe, Hamilton
Fournisseur: Hamilton
Pierce™ Blocker™ Casein in PBS, Thermo Scientific
Fournisseur: Thermo Scientific