MINI CAPSULE HALF SIZE 1.2UM PP CS5
Catalogue # 76497-420
Fournisseur: PARKER HANNIFIN TE
MINI CAPSULE HALF SIZE 1.2UM PP CS5
Catalogue # 76497-420
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 22-CHBPP-012-2
Frequently Bought Together
Leakbuster™ Specimen Containers
Fournisseur: Starplex
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Acetone ≥99.5% (by GC, corrected for water content), OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Fournisseur: Corning
Propanol-2 ≥70%, VWR Chemicals BDH®
Fournisseur: VWR International
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
GelRed® & GelGreen® Nucleic Acid Gel Stains, Biotium
Fournisseur: Biotium
Water for HPLC, Supelco®
Fournisseur: MilliporeSigma
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
B352 Specimen Bottles and Caps, Simport Scientific
Fournisseur: Simport Scientific
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
Anti-APP Mouse Monoclonal Antibody [clone: 9C4/Amyloid]
Fournisseur: Biolegend
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Sodium alginate, high viscosity
Fournisseur: Thermo Scientific Chemicals
LabKlenz™ 100 Alkaline Detergents
Fournisseur: Steris
Ozone AccuVac® Ampules, MR, Hach
Fournisseur: Hach
Calbiochem® Miracloth, EMD Millipore
Fournisseur: MilliporeSigma
MycoAlert™ Mycoplasma Detection Kits, Lonza
Fournisseur: LONZA PHARMA - BIOSCIENCE CA