CARTRIDGE 30IN 1UM BF/226 BAYONET SILCN
Catalogue # 76496-052
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 30IN 1UM BF/226 BAYONET SILCN
Catalogue # 76496-052
Fournisseur: PARKER HANNIFIN TE
Supplier Number: PCGF3-01C-PS
Frequently Bought Together
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
Parafilm® M Sealing Film
Fournisseur: Heathrow Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
Orion™ pH Buffer Bottles, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Acétone ≥99.5% ACS, VWR Chemicals BDH®
Fournisseur: VWR International
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
OneStep™ PCR Inhibitor Removal Kits, Zymo Research
Fournisseur: Zymo Research
SYRINGE STERILE WITHOUT NEEDLE 10ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Components and Accessories for Miele Lab Washers, Miele Professional
Fournisseur: Miele
COMPLEMENT GUINEA PIG NON-STERILE 10ML
Fournisseur: Rockland Immunochemical
BD P100 BLOOD COLLECTION TUBES (24/PK)
Fournisseur: BD
Potassium hydroxide 0.100 N (N/10) in isopropanol (IPA)
Fournisseur: Ricca Chemical
Circular Cover Glass, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC