CARTRIDGE 10IN 0.6UM BF/226 BAYONT SILCN
Catalogue # 76496-026
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 10IN 0.6UM BF/226 BAYONT SILCN
Catalogue # 76496-026
Fournisseur: PARKER HANNIFIN TE
Supplier Number: PCGF1-96C-PS
Frequently Bought Together
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
PREempt® Concentrate Disinfectants, Contec
Fournisseur: Contec
Poulten and Graf Fortuna™ Glass Syringes, Air-Tite Products
Fournisseur: Air-Tite
Preserved Starfish
Fournisseur: Avantor
VWR® Traceable® Conductivity Calibration Standards (CRM)
Fournisseur: VWR International
Acide chlorhydrique 1.0 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
VWR® Specimen Containers, PP with PE Caps
Fournisseur: VWR International
SingleQuots® EGM®-2 Endothelial Kit
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Components and Accessories for Miele Lab Washers, Miele Professional
Fournisseur: Miele
AGAR BRAIN HEART INFUSION 450 G
Fournisseur: BECTON DICKINSON DE MEXICO, S. MX
E. cloni® Electrocompetent Cells, Biosearch Technologies
Fournisseur: Lucigen
Phage Display Competent Cells, Biosearch Technologies
Fournisseur: Lucigen
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX