CARTRIDGE 30IN 0.45U PTFE BF/226 BAYONET
Catalogue # 76495-834
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 30IN 0.45U PTFE BF/226 BAYONET
Catalogue # 76495-834
Fournisseur: PARKER HANNIFIN TE
Supplier Number: ZCMT3-045C-LE
Frequently Bought Together
SYRINGE FOR INSULIN WITH NEEDLE 1ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Exel International® Insulin Syringes with Fixed Needles, Air-Tite Products
Fournisseur: Air-Tite
Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Fournisseur: Air-Tite
pH Test Strips, VWR Chemicals BDH®
Fournisseur: BDH
VWR® Multipurpose Containers, HDPE
Fournisseur: VWR International
Glucose Tolerance Drinks, Azer Scientific
Fournisseur: Azer Scientific
VWR® EZ-Vision® Dye-as-Loading Buffer, 6X
Fournisseur: VWR
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
COD Digestion vials, VWR Chemicals BDH®
Fournisseur: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
CL17 DPD Indicator, Hach
Fournisseur: Hach
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Fournisseur: TCI America
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Fournisseur: BD
ALKALINE CYANIDE REAGNT, 500ML
Fournisseur: Hach
Components and Accessories for Miele Lab Washers, Miele Professional
Fournisseur: Miele