CARTRIDGE 5IN 0.45UM PTFE MEMB 222/FLAT
Catalogue # 76495-746
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 5IN 0.45UM PTFE MEMB 222/FLAT
Catalogue # 76495-746
Fournisseur: PARKER HANNIFIN TE
Supplier Number: ZCMTK-045E-LE
Frequently Bought Together
Méthanol ≥99.9%, OmniSolv® for purge & trap, Supelco®
Fournisseur: MilliporeSigma
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
Quality Economy Brand Luer Slip Syringes, Air-Tite Products
Fournisseur: Air-Tite
MQuant® pH Test Strips, Supelco®,MilliporeSigma
Fournisseur: MilliporeSigma
Méthanol ≥99.8% ACS for HPLC, Supelco®
Fournisseur: MilliporeSigma
Leakbuster™ Specimen Containers
Fournisseur: Starplex
Hydrogen peroxide 3% (w/w) in aqueous solution stabilized, VWR Chemicals BDH®
Fournisseur: VWR International
MICROLITER® 700 Series Syringes, Hamilton
Fournisseur: Hamilton
B352 Specimen Bottles and Caps, Simport Scientific
Fournisseur: Simport Scientific
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Fournisseur: BD
Duke Standards™ Particle-Size Standards, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
SingleQuots® EGM®-2 Endothelial Kit
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Formaldehyde 8% in aqueous solution, EM Grade
Fournisseur: Electron Microscopy Sciences
Bovine Serum Albumin (BSA), Fraction V
Fournisseur: MilliporeSigma
Single-Use Needles, BD Medical
Fournisseur: BD
Reagent Sets for Colorimeters, Hach
Fournisseur: Hach
Acetonedimethyl acetal 98%
Fournisseur: Thermo Scientific Chemicals