CARTRIDGE 222/FLAT 20IN 0.65/0.2UM SILCN
Catalogue # 76495-030
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 222/FLAT 20IN 0.65/0.2UM SILCN
Catalogue # 76495-030
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10320-602-2-XL
Frequently Bought Together
Glycérol ≥99.7% ACS, Laboratory Reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Syringes and Tubing for MICROLAB® 500 Series Dispensers/Diluters, Hamilton
Fournisseur: Hamilton
Acide chlorhydrique 1.0 N analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Accessories for Micro-Osmometer, Advanced Instruments
Fournisseur: Advanced Instruments
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Masterflex® Hose Clamps, Acetal Copolymer, Avantor®
Fournisseur: VWR International
COD Digestion vials, VWR Chemicals BDH®
Fournisseur: VWR International
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Fournisseur: Cytiva
AGAR BRAIN HEART INFUSION 450 G
Fournisseur: BECTON DICKINSON DE MEXICO, S. MX
SYRINGE WITH LUER SLIP TIP STERILE 10ML
Fournisseur: BD
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
DAPT ≥98% (by NMR)
Fournisseur: Enzo Life Sciences
Human Recombinant CNTF (from <i>E. coli</i>)
Fournisseur: Peprotech
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Bovine Serum Albumin (BSA)
Fournisseur: Rockland Immunochemical
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Fournisseur: BD