CARTRIDGE PP DOE 10IN 0.65/0.2UM EPDM
Catalogue # 76494-998
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE PP DOE 10IN 0.65/0.2UM EPDM
Catalogue # 76494-998
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10110-602-1-XL
Frequently Bought Together
Orion™ Conductivity Standards, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Contrad® 70 Liquid Detergent
Fournisseur: Decon Labs
Orion™ pH Buffer Bottles, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Acide sulfurique 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Histoplex™ Histology Containers, Starplex®
Fournisseur: Starplex
SYRINGE STERILE WITHOUT NEEDLE 10ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Sodium chloride ≥99.0% ACS
Fournisseur: Thermo Scientific Chemicals
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Human Recombinant CNTF (from <i>E. coli</i>)
Fournisseur: Peprotech
Chemical Oxygen Demand (COD) Vials
Fournisseur: Chemetrics
Sodium octyl sulfate 99%
Fournisseur: Thermo Scientific Chemicals
Forget-Me-Not™ EvaGreen® qPCR Master Mixes, Biotium
Fournisseur: Biotium
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC