CARTRIDGE 222/FLAT 20IN 0.45/0.2UM SILCN
76494-970
Frequently Bought Together
Forceps, Stainless Steel, Sharp, Bent Form, Bürkle
: BURKLE INC
Ammonium sulfate ≥99.0% ACS, VWR Chemicals BDH®
: VWR International
Acétone ≥99.5% ACS, VWR Chemicals BDH®
: VWR International
Masterflex® Transfer Tubing, C-Flex®, Clear, Avantor®
: VWR International
Buffers, Reference Standards, Color-Coded, Ricca Chemical Company
: Ricca Chemical
GLUCOSE DRINK SIMPLY 75G CS24
: Azer Scientific
AGAR BRAIN HEART INFUSION 450 G
: BECTON DICKINSON DE MEXICO, S. MX
Scalpel Blades, Sterile, Mortech
: Mortech Manufacturing
Nunc® MicroWell™ 96-Well Plates, Thermo Scientific
: Thermo Fisher Scientific
STORAGE SOLUTION PH ELECTRODE
: Oakton
Sphingosine-1-phosphate ≥98% (by TLC)
: Enzo Life Sciences
Anti-Rabbit IgG H&L Goat Polyclonal Antibody (Cyanine 5)
: ANTIBODIES.COM LLC
Sol-M® Hypodermic Needles
: SOL-MILLENNIUM CANADA, INC. CA
di-Ammonium oxalate monohydraté, EMSURE® for analysis, Supelco®
: MilliporeSigma
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Amber
: DWK Life Sciences (KIMBLE)