CARTRIDGE 222/FLAT 20IN 0.45/0.2UM EPDM
Catalogue # 76494-964
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 222/FLAT 20IN 0.45/0.2UM EPDM
Catalogue # 76494-964
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10320-402-1-XP
Frequently Bought Together
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Syringes and Tubing for MICROLAB® 500 Series Dispensers/Diluters, Hamilton
Fournisseur: Hamilton
Potassium dihydrogen phosphate ≥99% (by acidimetry), GR ACS, Supelco®
Fournisseur: MilliporeSigma
Difco™ Granulated Agar, BD Biosciences
Fournisseur: BD
Bacto™ Brain Heart Infusion, BD Diagnostics
Fournisseur: BD
Kerosene, Supelco®
Fournisseur: MilliporeSigma
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
Zymoclean™ Gel DNA Recovery Kits, Zymo Research
Fournisseur: Zymo Research
Preserved Starfish
Fournisseur: Avantor
Single-Use Needles, BD Medical
Fournisseur: BD
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
ACETIC RINSE SOLUTION 4L
Fournisseur: Ostrem Chemical
SYRINGE WITH NEEDLE 5ML
Fournisseur: AMX INDUSTRIAL TRADE MX
Phage Display Competent Cells, Biosearch Technologies
Fournisseur: Lucigen
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Silica gel, BAKER ANALYZED® ACS (6-18 Mesh) desiccant, with moisture indicator, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
MOUNTING MEDIUM WITH DAPI 100 TESTS
Fournisseur: Enzo Life Sciences
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Fournisseur: BD
E. cloni® Electrocompetent Cells, Biosearch Technologies
Fournisseur: Lucigen
Dri-Contrad® Powder Detergent
Fournisseur: Decon Labs