CARTRIDGE DOE 20IN 0.45/0.2UM SILICONE
Catalogue # 76494-946
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE DOE 20IN 0.45/0.2UM SILICONE
Catalogue # 76494-946
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10120-402-2-XP
Frequently Bought Together
VWR® Specimen Containers with Dual Click
Fournisseur: VWR International
Sodium hydroxyde 0.5 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
Eau ASTM Type II, VWR Chemicals BDH®
Fournisseur: VWR International
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
Residual Solvents Class 2 - Mix A (15 components), Restek
Fournisseur: Restek
Sodium Iodide
Fournisseur: Ward's Science
Chloroforme, purified, Macron Fine Chemicals™
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
Sodium chloride ≥99.0% ACS
Fournisseur: Thermo Scientific Chemicals
Sol-Care® Safety Winged Blood Collection Needles, Sol M
Fournisseur: SOL-MILLENNIUM CANADA, INC. CA
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Warthin-Starry silver plating kit for histology
Fournisseur: Azer Scientific Inc.
Specimen Shipping Boxes for Ambient Transport, Therapak®
Fournisseur: Therapak, LLC
Griffin Style Beakers, Mortech
Fournisseur: Mortech Manufacturing
Water Quality Meter, Sper Scientific
Fournisseur: Sper Scientific
Sodium-tert-butanolate 97%
Fournisseur: Thermo Scientific Chemicals
Skim Milk Powder, CRITERION™, Hardy Diagnostics
Fournisseur: HARDY DIAGNOSTICS CA
PREP SOLUTION 30ML
Fournisseur: Zymo Research
Anisole 99%
Fournisseur: Thermo Scientific Chemicals