CARTRIDGE 222/FIN 10IN 0.65UM SILICONE
Catalog # 76494-914
Supplier: PARKER HANNIFIN TE
CARTRIDGE 222/FIN 10IN 0.65UM SILICONE
Catalog # 76494-914
Supplier: PARKER HANNIFIN TE
Supplier Number: 25-10810-006-2-B
Frequently Bought Together
Masterflex® Luer Stopcock Fittings, Polycarbonate, Avantor®
Supplier: VWR International
VWR® ClikSeal Specimen Containers
Supplier: VWR International
Sodium hydroxide 5 N in aqueous solution, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Premium Aluminum Foil
Supplier: VWR International
Propanol-2 ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Tetrahydrofuran ≥99.9% (by GC) unstabilised, OmniSolv®, Supelco®
Supplier: MilliporeSigma
VACUETTE® Heparin Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Tryptone to 2xYT Broth, Life Technologies
Supplier: BD
VACUETTE® Secondary Tubes, Greiner Bio-One
Supplier: Greiner Bio-One
Difco™ Granulated Agar, BD Biosciences
Supplier: BD
VWR® Weighing Paper
Supplier: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Alcool ≥70%, Reagent Grade (denatured with methanol and isopropanol), VWR Chemicals BDH®
Supplier: VWR International
High Sensitivity RNA ScreenTape Analysis
Supplier: AGILENT TECHNOLOGIES (GENOMICS) CA
Sodium stearyl fumarate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Fetal bovine serum (FBS), premium grade, Corning®
Supplier: Corning
E.Z.N.A.® Total RNA Kit I, Omega Bio-tek®
Supplier: Omega Bio-Tek
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo