CARTRIDGE 226/FIN 5IN 0.2UM SILICONE
Catalog # 76494-838
Supplier: PARKER HANNIFIN TE
CARTRIDGE 226/FIN 5IN 0.2UM SILICONE
Catalog # 76494-838
Supplier: PARKER HANNIFIN TE
Supplier Number: 25-10705-002-2-B
Frequently Bought Together
Accessories for Multi-Sample Osmometer, Model 3900, Advanced Instruments
Supplier: Advanced Instruments
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Leakbuster™ Specimen Containers
Supplier: Starplex
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
Supplier: Thermo Scientific
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Supplier: MEDONE SURGICAL, INC
Starch 1% (w/v) in aqueous solution for iodometric titrations mercury-free
Supplier: Ricca Chemical
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
SingleQuots® EGM®-2 Endothelial Kit
Supplier: LONZA PHARMA - BIOSCIENCE CA
SYR ULTRAFINE II 30G 1CC PK100
Supplier: Cardinal Health
Disposable Base Mold, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
DNA MINICOLUMNS V-SPIN HIBIND PK1000
Supplier: Omega Bio-Tek
Flow Cell Priming Kit
Supplier: Oxford Nanopore Technologies
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo
TOTAL CHLORINE IND SOL 473ML
Supplier: Hach
Dextrose, anhydrous 97.5-102.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC