CARTRIDGE PP 222/FLAT 20IN 0.1UM EPDM
Catalogue # 76494-748
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE PP 222/FLAT 20IN 0.1UM EPDM
Catalogue # 76494-748
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10320-001-1-B
Frequently Bought Together
VWR® Multipurpose Containers, HDPE
Fournisseur: VWR International
Formalin, VWRClick, neutral buffered prefill
Fournisseur: VWR International
Water, distilled
Fournisseur: GROWCELLS
Glucose Tolerance Drinks, Azer Scientific
Fournisseur: Azer Scientific
VWR® Ultra Pure Grade Phosphate Buffered Saline
Fournisseur: VWR
Ferric chloride 0.025% (w/v) in aqueous solution
Fournisseur: Ricca Chemical
PerfeCTa® qPCR ToughMix®, QuantaBio
Fournisseur: Quantabio
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Corning® Matrigel® Basement Membrane Matrix, Corning
Fournisseur: Corning
Alcool, anhydrous 94.0-96.0% (v/v), BAKER ANANYZED™ ACS (denatured with methanol and isopropanol), J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
SingleQuots® EGM®-2 Endothelial Kit
Fournisseur: LONZA PHARMA - BIOSCIENCE CA
Staining Dish Set for 30 Slides, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Iodomethane ≥99.5% stabilized
Fournisseur: Thermo Scientific Chemicals
Orion™ Fluoride Electrodes, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
Salicylaldehyde azine 97%
Fournisseur: Thermo Scientific Chemicals
Tryptone to 2xYT Broth, Life Technologies
Fournisseur: BD