CARTRIDGE PP 222/FLAT 5IN 0.2UM EPDM
Catalogue # 76494-736
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE PP 222/FLAT 5IN 0.2UM EPDM
Catalogue # 76494-736
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 25-10305-002-1-B
Frequently Bought Together
VWR® Premium Aluminum Foil
Fournisseur: VWR International
Single-Use Needles, BD Medical
Fournisseur: BD
Éthanol dénaturé, anhydrous
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Fournisseur: Air-Tite
Potassium hydroxide ACS, VWR Chemicals BDH®
Fournisseur: VWR International
Orion™ pH Electrode Cleaning and Storage Solutions, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Glucose Tolerance Drinks, Azer Scientific
Fournisseur: Azer Scientific
BD GasPak™ EZ Container Systems, BD Diagnostics
Fournisseur: BD
Ocean Pacific® Ninja™ Powder-Free Nitrile Medical Examination Gloves, Medicom
Fournisseur: Medicom
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Sodium hydroxyde 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
BD Syringes only with Luer-Lok™, Slip or Eccentric tips
Fournisseur: BD
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
Hydrochloric acid 34 - 37%, OmniTrace® for trace metal analysis, Supelco®
Fournisseur: MilliporeSigma
Corning® Fetal Bovine Serum, Regular, CA sourced, Corning
Fournisseur: Corning
E. cloni® Electrocompetent Cells, Biosearch Technologies
Fournisseur: Lucigen
Calcium sulfate, anhydrous 99%
Fournisseur: Thermo Scientific Chemicals
Aluminium fluoride, anhydrous ≥99.99% (metals basis)
Fournisseur: Thermo Scientific Chemicals