CARTRIDGE 226/FLAT 10IN 0.65UM EPDM
Catalog # 76494-676
Supplier: PARKER HANNIFIN TE
CARTRIDGE 226/FLAT 10IN 0.65UM EPDM
Catalog # 76494-676
Supplier: PARKER HANNIFIN TE
Supplier Number: 25-10210-006-1-B
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
BD Vacutainer® Venous Blood Collection Tubes
Supplier: BD
Orion™ pH Buffer Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Water for HPLC, Supelco®
Supplier: MilliporeSigma
Self Standing Sample Tubes, 5 and 10 ml, Globe Scientific
Supplier: Globe Scientific
Dichlorométhane ≥99.5% (by GC), GR ACS ACS, Supelco®
Supplier: MilliporeSigma
Water USP for injection (WFI), sterile
Supplier: Cytiva
di-Potassium hydrogen phosphate, dried 98+%
Supplier: Thermo Scientific Chemicals
pH Buffer Solutions, Hach
Supplier: Hach
Formic acid ≥96% ACS
Supplier: Thermo Scientific Chemicals
E. cloni® Electrocompetent Cells, Biosearch Technologies
Supplier: Lucigen
Acetone, low water content ≥99.3%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
2-Deoxy-D-glucose 98%
Supplier: Thermo Scientific Chemicals
BD P100 BLOOD COLLECTION TUBES (24/PK)
Supplier: BD
Technical Buffer Solutions, Sachets
Supplier: Mettler Toledo