CARTRIDGE 222/FIN 30IN 1UM SILICONE 5 MM
Catalogue # 76494-660
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE 222/FIN 30IN 1UM SILICONE 5 MM
Catalogue # 76494-660
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 28-19830-010-21
Frequently Bought Together
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Fournisseur: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Hexane (mixture of isomers) ≥98.5%, OmniSolv®, Supelco®
Fournisseur: MilliporeSigma
Potassium hydrogen carbonate, Reagent Grade
Fournisseur: VWR
BD SafetyGlide™ Syringes for Insulin, TB, and Allergy, BD
Fournisseur: BD
Trypan blue solution 0,4% (w/v) in PBS stain for histology pH 7.5 ±0.5, Corning®
Fournisseur: Corning
Corning® Trace Elements C, Corning
Fournisseur: Corning
LabKlenz™ 100 Alkaline Detergents
Fournisseur: Steris
VWR® Disposable Scalpels with Measurements
Fournisseur: VWR International
Conflikt® Ready-to-use Disinfectant
Fournisseur: Decon Labs
VeriSpec® Molybdenum Standard for ICP 1000 ppm in 4% NH₃, Ricca Chemical Company
Fournisseur: Ricca Chemical
Dimethyl sulfoxide ≥99.9% for HPLC
Fournisseur: Thermo Scientific Chemicals
NitriVer 3 TNT Reagent Set, Nitrogen-Nitrite, Low Range, Hach
Fournisseur: Hach