CARTRIDGE 226/FLAT 30IN 0.25UM SILCN 5MM
76494-416
Frequently Bought Together
VWR® Self-Seal Sterilization Pouches
: VWR International
2,2,4-Triméthylpentane ≥99.0% ACS
: AVANTOR PERFORMANCE MATERIAL LLC
VWR® Weighing Paper
: VWR International
Contrad® 70 Liquid Detergent
: Decon Labs
Sodium hydroxide 0.100 N (N/10) in aqueous solution
: Ricca Chemical
VWR® Reagent alcohol
: VWR International
RNase AWAY® and DNA AWAY™ Surface Decontaminants, Molecular BioProducts
: Thermo Scientific
Histoplex™ Histology Containers, Starplex®
: Starplex
SYRINGE STERILE WITHOUT NEEDLE 10ML
: AMX INDUSTRIAL TRADE MX
Diversey™ Oxivir® Tb Disinfectant Solution, Antylia Scientific
: Antyila Scientific
Hydrochloric acid 3.00 N
: Ricca Chemical
LabKlenz™ 100 Alkaline Detergents
: Steris
Laboratory pH Sensors, Hamilton Company
: Hamilton