CARTRIDGE PP DOE 20IN 3UM EPDM 5 MM
Catalogue # 76494-292
Fournisseur: PARKER HANNIFIN TE
CARTRIDGE PP DOE 20IN 3UM EPDM 5 MM
Catalogue # 76494-292
Fournisseur: PARKER HANNIFIN TE
Supplier Number: 28-19120-030-11
Frequently Bought Together
Air-Tite Vet Premium Hypodermic Needles, Air-Tite Products
Fournisseur: Air-Tite
Target™ All-Plastic Disposable Luer-Lok Syringes, Thermo Scientific
Fournisseur: Thermo Scientific
Argent nitrate 0.1 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Fournisseur: VWR International
INCYTO C-Chip™ Disposable Hemacytometers, SKC
Fournisseur: MEDONE SURGICAL, INC
Formalin 10%, neutral buffered, VWR®
Fournisseur: VWR International
BD Vacutainer® Venous Blood Collection Tubes
Fournisseur: BD
MiniCollect® Serum and Plasma Tubes, Greiner Bio-One
Fournisseur: Greiner Bio-One
Ammonia CHEMets and VACUettes High Range Visual Kits, CHEMetrics
Fournisseur: Chemetrics
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
HSW® Norm-Ject® Sterile Luer-Slip Syringes, Air-Tite
Fournisseur: Air-Tite
pH Reference Standard Buffers, VWR Chemicals BDH®
Fournisseur: BDH
Phenols, CHEMets Visual Kit, CHEMetrics
Fournisseur: CHEMetrics
Acetonitrile ≥99.9% (by GC), OmniSolv®, gradient grade for HPLC, Supelco®
Fournisseur: MilliporeSigma
High Sensitivity RNA ScreenTape Analysis
Fournisseur: AGILENT TECHNOLOGIES (GENOMICS) CA
HydroKlean Solution, Contec®
Fournisseur: Contec
CL17 DPD Indicator, Hach
Fournisseur: Hach
Acide acétique glacial ≥99% USP, Multi-Compendial for synthesis, for biochemistry, J.T.Baker®
Fournisseur: AVANTOR PERFORMANCE MATERIAL LLC