CARTRIDGE PP DOE STD 5IN 1UM EPDM 5 MM
Catalog # 76494-270
Supplier: PARKER HANNIFIN TE
CARTRIDGE PP DOE STD 5IN 1UM EPDM 5 MM
Catalog # 76494-270
Supplier: PARKER HANNIFIN TE
Supplier Number: 28-19105-010-11
Frequently Bought Together
Leakbuster™ Specimen Containers
Supplier: Starplex
Convertors® Paper/Film Gas/Steam Sterilization Pouches, Cardinal Health®
Supplier: Cardinal Health
Micro Check Chlorine Test Strips
Supplier: HF Scientific
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
B352 Specimen Bottles and Caps, Simport Scientific
Supplier: Simport Scientific
DIBMA Screening Kit, HEPES
Supplier: CUBE BIOTECH
DAPT ≥98% (by NMR)
Supplier: Enzo Life Sciences
Ward's® Live <i>Bacillus subtilis</i> Culture
Supplier: Avantor
Potassium stannate trihydrate ≥95% (metals basis)
Supplier: Thermo Scientific Chemicals
Formaldehyde 10% (v/v) in aqueous solution histological fixative, (non-buffered)
Supplier: Ricca Chemical
Trypan blue 0.4% in PBS
Supplier: Cytiva
Sulfuric acid 0.1 N concentrated aqueous solution, Titrisol®, Supelco®
Supplier: MilliporeSigma
Puromycin dihydrochloride ≥98% (by HPLC)
Supplier: Enzo Life Sciences
Reagent Sets for Colorimeters, Hach
Supplier: Hach
Potassium hydroxide 0.500 N (N/2) in denatured alcohol
Supplier: Ricca Chemical
OSOM® Ultra Strep A Test Kits, Sekisui Diagnostics
Supplier: Sekisui Diagnostics