InocuLoop Inoculating Loops, Caplugs
Needle, PS, Green
  76492-646
 :   Caplugs
 :  333-5013-G1I
InocuLoop Inoculating Loops, Caplugs
  76492-646
 :  Caplugs
InocuLoop Inoculating Loops, Caplugs
InocuLoop Inoculating Loops, Caplugs
Needle, PS, Green
$305.45
Carton of 500
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
  • Made of flexible plastic for easy spreading
  • Can also be used as a picker for isolating colonies
  • Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
  • Color:
    Green
  • Désignation:
    Needle
  • Matière:
    PS