InocuLoop Inoculating Loops, Caplugs
Needle, PS, Green
76492-646
: Caplugs
: 333-5013-G1I
Inoculoop disposable inoculating loops have patented four-sided spreaders, enabling the inoculation of a petri dish with one, not multiple, disposable loops. Make bacterial inoculation and quantitative procedures faster and more efficient.
- Made of flexible plastic for easy spreading
- Can also be used as a picker for isolating colonies
- Pre-sterilized Inoculoops are gamma-irritated
Suitable in anaerobic chambers, in the field and even in satellite metal loops.
- Color:Green
- Désignation:Needle
- Matière:PS